Split-hinged tube furnaces, for horizontal operation, HST and HZS
Supplier: CARBOLITE
HST12200301 HST12400301 HST12600301 HST12900301 HST122003216P1 HST122003508P1 HST12300301 HST123003216P1 HST123003508P1 HST124003216P1 HST124003508P1 HST126003216P1 HST126003508P1 HST129003216P1 HST129003508P1 HZS12600301 HZS126003216P1 HZS126003508P1 HZS12900301 HZS129003216P1 HZS129003508P1 HST120200230SN HST120300230SN HST120400230SN HST120600230SN HST120900230SN HZS120900230SN
455-3161EA 5930 EUR
455-3161 455-3162 455-3163 455-3164 CARBHST122003216P1 CARBHST122003508P1 CARBHST12300301 CARBHST123003216P1 CARBHST123003508P1 CARBHST124003216P1 CARBHST124003508P1 CARBHST126003216P1 CARBHST126003508P1 CARBHST129003216P1 CARBHST129003508P1 CARBHZS12600301 CARBHZS126003216P1 CARBHZS126003508P1 CARBHZS12900301 CARBHZS129003216P1 CARBHZS129003508P1 CARBHST120200230SN CARBHST120300230SN CARBHST120400230SN CARBHST120600230SN CARBHST120900230SN CARBHZS120900230SN
Split-hinged tube furnaces, for horizontal operation, HST and HZS
Ovens Tube Furnaces
The horizontal split tube furnace range HST (single zone) and HZS (three zone) uses free radiating wire elements embedded within the insulation of the furnace body. The benefit of this design is its flexibility; with the use of tube adapters the same furnace can be used with a variety of tube diameters. These furnaces comprise a furnace body which is hinged and split into two halves along its length. This makes exchange of work tubes easier and also enables the tube furnace to be used with reactors or work tubes where end flanges would make insertion into a non-split furnace difficult.
Order Now
SPECIFICATIONS
Environmentally Preferable