Large bore tube furnaces, GHA and GHC
Supplier: CARBOLITE
GHA12/450 GHC126003216P1 CGHA12600301 GHC121200301 GHA12/450/301 GHA120600230SN GHA120450230SN GHC120600230SN
CARBGHA12/450EA 0 EUR
CARBGHA12/450 CARBGHC126003216P1 CARBCGHA12600301 CARBGHC121200301 CARBGHA12/450/301 CARBGHA120600230SN CARBGHA120450230SN CARBGHC120600230SN
Large bore tube furnaces, GHA and GHC
Ovens Tube Furnaces
The modular horizontal tube furnace range GHA (single zone) and GHC (3-zone) uses free radiating wire elements embedded within the insulation of the furnace body. The benefit of this design is its flexibility; with the use of tube adapters the same furnace can be used with a variety of tube diameters.
  • Maximum operating temperature 1200 °C
  • Carbolite Gero 301 controller, with single ramp to set-point & process timer
  • Accepts work tubes with outer diameter up to 170 mm
  • Long life, rapid heating, resistance wire elements mounted in rigid, vacuum formed
More Product Information